Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028049.2 | NP_082325.2 | 5, 6 | 734 | ||
GenBank mRNA | AK002745.1 | - | 5, 6 | 614 | ||
AK005473.1 | - | 4, 5 | 562 | |||
AK182889.1 | - | 1, 2 | 169 | |||
AK207864.1 | - | 1, 2 | 169 | |||
AK211102.1 | - | 1, 2 | 36 | |||
AK213133.1 | - | 1, 2 | 169 | |||
AK219248.1 | - | 1, 2 | 169 | |||
BC018273.1 | - | 5, 6 | 674 | 4458296 |