Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030081.2 | NP_084357.2 | 3 | 495 | ||
GenBank mRNA | AK019909.2 | - | 3 | 534 | ||
BC017622.1 | - | 3 | 542 | |||
BC031135.1 | - | 2 | 326 | 4971294 | ||
BC049106.1 | - | 3 | 495 | 6412839 |