Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002744.4 | NP_002735.3 | 3, 4 | 437 | ||
XM_011541773.1 | XP_011540075.1 | 3, 4 | 433 | |||
XM_011541775.2 | XP_011540077.1 | 3, 4 | 433 | |||
XM_017001789.1 | XP_016857278.1 | 3, 4 | 433 | |||
XM_017001790.1 | XP_016857279.1 | 3, 4 | 433 | |||
XM_017001803.1 | XP_016857292.1 | 3, 4 | 433 | |||
GenBank mRNA | AK290995.1 | - | 3, 4 | 276 | ||
BC008058.2 | - | 3, 4 | 384 | 2987996 | ||
BC014270.2 | - | 3, 4 | 437 | 3835020 | ||
BT007082.1 | - | 3, 4 | 276 | |||
L14283.1 | - | 3, 4 | 276 | |||
Z15108.1 | - | 3, 4 | 258 |