Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033268.4 | NP_150371.4 | 18, 19 | 2529 | ||
GenBank mRNA | AK003570.1 | - | 2, 3 | 292 | ||
AY036877.1 | - | 19, 20 | 2473 | |||
BC089579.1 | - | 18, 19 | 2529 | 6757718 |