Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053103.5 | NP_444333.3 | 4 | 556 | ||
GenBank mRNA | AK007446.1 | - | 4 | 428 | ||
AK081564.1 | - | 4 | 414 | |||
AK147401.1 | - | 4 | 421 | |||
AK166220.1 | - | 4 | 406 | |||
AK170546.1 | - | 4 | 574 | |||
BC137709.1 | - | 3 | 342 | 8860728 | ||
BC137717.1 | - | 3 | 342 | 8860736 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF288221.1 | - | 460 | Chr. - - [-] on Build |