Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053267.2 | NP_444497.1 | 5 | 666 | ||
GenBank mRNA | AK018791.1 | - | 5 | 502 | ||
AK035312.1 | - | 5 | 456 | |||
AK046034.1 | - | 5 | 458 | |||
AK079358.1 | - | 5 | 485 | |||
AK079576.1 | - | 2 | 1276 | |||
AY043488.1 | - | 5 | 482 | |||
BC019742.1 | - | 5 | 358 | |||
BC147332.1 | - | 5 | 576 | 9056406 | ||
BC147333.1 | - | 5 | 578 | 9056396 |