Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxfp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289564.1 | NP_001276493.1 | 4, 5 | 560 | ||
NM_001289566.1 | NP_001276495.1 | 4, 5 | 560 | |||
NM_080468.2 | NP_536716.2 | 4, 5 | 560 | |||
GenBank mRNA | AF346501.1 | - | 4, 5 | 560 | ||
BC120741.1 | - | 4, 5 | 529 | 40129664 | ||
BC144802.1 | - | 4, 5 | 529 | 9053350 | ||
BC144803.1 | - | 4, 5 | 529 | 9053351 |