Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133726.2 | NP_598487.1 | 3 | 296 | ||
GenBank mRNA | AK013981.1 | - | 3 | 283 | ||
AK146528.1 | - | 3 | 292 | |||
AK147169.1 | - | 319 | ||||
AK152270.1 | - | 3 | 292 | |||
AK154658.1 | - | 285 | ||||
AK159149.1 | - | 276 | ||||
AK205921.1 | - | 2 | 53 | |||
BC003843.1 | - | 3 | 296 | 3495262 | ||
CT010318.1 | - | 3 | 196 |