Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164743.1 | NP_001158215.1 | 9 | 1386 | ||
NM_134069.3 | NP_598830.2 | 10 | 1510 | |||
GenBank mRNA | AK128896.1 | - | 10 | 1494 | ||
AK143896.1 | - | 10 | 1482 | |||
BC018252.1 | - | 10 | 1439 | 4240254 | ||
BC024459.1 | - | 10 | 1466 | 4975341 | ||
BC034179.1 | - | 10 | 1330 | 4972500 | ||
BC034183.1 | - | 10 | 1315 | 4973620 |