Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144898.2 | NP_659147.2 | 8, 9 | 856 | ||
GenBank mRNA | AK134001.1 | - | 313 | |||
AK139708.1 | - | 835 | ||||
AK163280.1 | - | 8, 9 | 716 | |||
AK166236.1 | - | 8, 9 | 828 | |||
AK166834.1 | - | 1, 2 | 2380 | |||
AK167586.1 | - | 8, 9 | 804 | |||
AK170160.1 | - | 8, 9 | 839 | |||
BC008103.1 | - | 8, 9 | 806 | 3966815 | ||
BC108354.1 | - | 8, 9 | 790 | 5035560 |