Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cetn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145825.2 | NP_665824.1 | 3 | 321 | ||
GenBank mRNA | AF362367.1 | - | 3 | 321 | ||
AK078275.1 | - | 2 | 2236 | |||
AK131808.1 | - | 3 | 309 | |||
AK166515.1 | - | 2 | 237 | |||
BC038154.1 | - | 2 | 212 | |||
BC060991.1 | - | 3 | 273 | 6742836 | ||
BC087905.1 | - | 3 | 299 | 6707456 |