Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6kl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146244.4 | NP_666356.2 | 11 | 2380 | ||
GenBank mRNA | AK031847.1 | - | 11 | 2375 | ||
AK036734.1 | - | 5 | 1918 | |||
AK044050.1 | - | 11 | 2371 | |||
BC024798.1 | - | 9 | 2194 | 5358742 | ||
BC027298.1 | - | 11 | 2334 | 3672127 | ||
BY651033.1 | - | 1 | 265 |