Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152813.3 | NP_690026.2 | 15 | 2952 | ||
GenBank mRNA | AK011892.1 | - | 11 | 1883 | ||
AK046669.1 | - | 15 | 2918 | |||
AK144950.1 | - | 3990 | ||||
AK173311.1 | - | 13 | 4910 | |||
BC031392.1 | - | 15 | 2896 | 4952407 | ||
BY589869.1 | - | 1 | 371 |