Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cadm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153112.3 | NP_694752.1 | 9 | 1478 | ||
GenBank mRNA | AK143204.1 | - | 9 | 1479 | ||
AY059394.1 | - | 9 | 1402 |