Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172136.1 | NP_001165607.1 | 7 | 971 | ||
NM_172456.3 | NP_766044.1 | 6 | 936 | |||
GenBank mRNA | AK028887.1 | - | 6 | 900 | ||
AK034654.1 | - | 7 | 933 | |||
AK051300.1 | - | 7 | 935 | |||
AK148426.1 | - | 6 | 917 | |||
AK162566.1 | - | 6 | 899 | |||
AK163555.1 | - | 6 | 900 | |||
AK184683.1 | - | 1 | 144 | |||
BC052171.1 | - | 5 | 742 | |||
BC059920.1 | - | 7 | 914 | |||
BC064730.1 | - | 7 | 986 | |||
BC094589.1 | - | 4 | 3422 | |||
BC119024.1 | - | 6 | 900 | 8733773 |