Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175358.4 | NP_780567.1 | 2, 3 | 427 | ||
GenBank mRNA | AK029137.1 | - | 2, 3 | 167 | ||
AK032922.1 | - | 2, 3 | 148 | |||
AK077949.1 | - | 2, 3 | 176 | |||
AK132304.1 | - | 2, 3 | 428 | |||
AK160360.1 | - | 2, 3 | 167 |