Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smcr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085440.1 | NP_001078909.1 | 1 | 1166 | ||
NM_175491.4 | NP_780700.1 | 1 | 1166 | |||
GenBank mRNA | AK083739.1 | - | 1 | 1150 | ||
AK145010.1 | - | 1 | 1152 | |||
AK164045.1 | - | 1 | 1155 | |||
BC085095.1 | - | 1 | 796 | 30732606 |