Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285928.1 | NP_001272857.1 | 4 | 591 | ||
NM_001285929.1 | NP_001272858.1 | 3 | 480 | |||
NM_001285930.1 | NP_001272859.1 | 3 | 344 | |||
NM_178671.5 | NP_848786.1 | 2 | 213 | |||
GenBank mRNA | AK045964.1 | - | 2 | 171 | ||
AK046949.1 | - | 4 | 592 | |||
AK077656.1 | - | 2 | 215 | |||
AK080662.1 | - | 2 | 179 | |||
AK133185.1 | - | 2 | 199 | |||
AK161245.1 | - | 2 | 200 | |||
AK161296.1 | - | 2 | 176 | |||
BC027099.1 | - | 3 | 344 | 5041000 |