Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178710.3 | NP_848825.2 | 7 | 988 | ||
GenBank mRNA | AB067780.1 | - | 7 | 889 | ||
AK090386.1 | - | 7 | 889 |