Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrfn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178714.4 | NP_848829.2 | 3 | 709 | ||
GenBank mRNA | AK034245.1 | - | 3 | 710 | ||
AK048443.1 | - | 3 | 795 | |||
AK083920.1 | - | 3 | 1904 | |||
AK220546.1 | - | 3 | 1393 | |||
BC052038.1 | - | 3 | 539 | 5708346 |