Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnpepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181405.4 | NP_852070.3 | 4 | 1005 | ||
GenBank mRNA | AK154881.1 | - | 4 | 1006 | ||
AK183521.1 | - | 2 | 86 | |||
BC031789.1 | - | 4 | 425 | 4006916 | ||
BQ559332.1 | - | 2 | 189 | |||
CA315475.1 | - | 4 | 699 |