Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183390.3 | NP_899246.2 | 3 | 932 | ||
GenBank mRNA | AY350736.1 | - | 3 | 860 | ||
BC100568.1 | - | 3 | 919 | 30073940 |