Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005448.2 | NP_005439.2 | 2 | 433 | ||
GenBank mRNA | BC069155.1 | - | 2 | 384 | 7217015 | |
BC117264.1 | - | 2 | 433 | 40125815 | ||
BC117266.1 | - | 2 | 433 | 40125817 |