Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282236.1 | NP_001269165.1 | 9 | 1798 | ||
NM_004196.4 | NP_004187.2 | 8 | 946 | |||
XM_005268157.2 | XP_005268214.1 | 9 | 1538 | |||
XM_005268160.4 | XP_005268217.1 | 6 | 2634 | |||
XM_011537276.1 | XP_011535578.1 | 9 | 948 | |||
XM_017021729.1 | XP_016877218.1 | 9 | 1430 | |||
XM_017021732.1 | XP_016877221.1 | 9 | 1517 | |||
GenBank mRNA | AK308732.1 | - | 9 | 1793 | ||
BC104977.1 | - | 8 | 921 | 8143980 | ||
BX099583.1 | - | 1 | 48 | |||
X66358.1 | - | 8 | 1106 |