Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lzts1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199364.2 | NP_955396.2 | 1 | 107 | ||
GenBank mRNA | AF288601.1 | - | 1 | 107 | ||
BC127606.1 | - | 1 | 106 | |||
BC127607.1 | - | 1 | 107 | 40057969 | ||
BC139350.1 | - | 1 | 163 | 8862372 | ||
BC139352.1 | - | 1 | 163 | 8862374 |