Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203507.3 | NP_987103.1 | 8 | 1048 | ||
GenBank mRNA | AK004597.1 | - | 8 | 1002 | ||
AK016505.1 | - | 8 | 1022 | |||
AK038266.1 | - | 1 | 477 | |||
BC016198.1 | - | 8 | 984 | 4503404 |