Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMLD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177465.2 | NP_001170936.1 | 5 | 3156 | ||
NM_001177466.2 | NP_001170937.1 | 6 | 3270 | |||
NM_005491.4 | NP_005482.2 | 7 | 3345 | |||
XM_006724802.3 | XP_006724865.1 | 9 | 3693 | |||
XM_006724803.3 | XP_006724866.1 | 8 | 3618 | |||
XM_011531092.2 | XP_011529394.1 | 7 | 3326 | |||
XM_011531093.2 | XP_011529395.1 | 8 | 3456 | |||
XM_011531095.2 | XP_011529397.1 | 8 | 3570 | |||
XM_017029188.1 | XP_016884677.1 | 9 | 3565 | |||
XM_017029189.1 | XP_016884678.1 | 8 | 3443 | |||
GenBank mRNA | AK094563.1 | - | 2 | 1635 | ||
AK125184.1 | - | 2 | 2665 | |||
AK294478.1 | - | 5 | 3015 | |||
BX537560.1 | - | 2 | 5016 | |||
U46023.1 | - | 7 | 3346 |