Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008914.2 | NP_032940.1 | 14 | 2224 | ||
GenBank mRNA | AK037615.1 | - | 1 | 877 | ||
BC037668.1 | - | 14 | 2047 | |||
BC049860.1 | - | 1 | 1199 | |||
BC066000.1 | - | 14 | 2196 | 5699179 |