Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkar2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011158.3 | NP_035288.2 | 3, 4 | 594 | ||
GenBank mRNA | AK011647.1 | - | 3, 4 | 552 | ||
AK041013.1 | - | 3, 4 | 568 | |||
AK081805.1 | - | 3, 4 | 371 | |||
AK135621.1 | - | 3, 4 | 596 | |||
AK138963.1 | - | 3, 4 | 560 | |||
BC048710.1 | - | 3, 4 | 550 | 6506827 |