Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013921.3 | NP_038949.2 | 6 | 1126 | ||
GenBank mRNA | AJ243866.1 | - | 6 | 1089 | ||
AK008263.1 | - | 5 | 971 | |||
BC010970.1 | - | 6 | 1119 | 4216644 | ||
BC040348.1 | - | 6 | 1126 | 4206523 |