Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163354.1 | NP_001156826.1 | 6 | 524 | ||
NM_001163355.1 | NP_001156827.1 | 6 | 524 | |||
NM_021536.7 | NP_067511.4 | 6 | 524 | |||
GenBank mRNA | AK019059.1 | - | 6 | 484 | ||
AK028597.1 | - | 6 | 467 | |||
AK031139.1 | - | 4 | 186 | |||
AK032096.1 | - | 6 | 497 | |||
AK041119.1 | - | 6 | 460 | |||
AK054472.1 | - | 6 | 495 | |||
AK078955.1 | - | 6 | 467 | |||
AK138830.1 | - | 6 | 465 | |||
AK141621.1 | - | 6 | 351 | |||
AK147749.1 | - | 6 | 503 | |||
AK147801.1 | - | 6 | 529 | |||
AK167502.1 | - | 6 | 524 | |||
AK169811.1 | - | 6 | 493 | |||
AK191854.1 | - | 4 | 158 | |||
BC046785.1 | - | 6 | 507 | 6416127 | ||
BC058350.1 | - | 6 | 439 | 6815832 |