Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif1ax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090053.1 | - | 2739 | |||
AK134827.1 | - | 2693 | ||||
AK134969.1 | - | 2693 | ||||
AK136814.1 | - | 1331 | ||||
AK161887.1 | - | 691 | ||||
AK165574.1 | - | 2735 |