Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cand2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025958.2 | NP_080234.2 | 6 | 875 | ||
GenBank mRNA | AK129186.1 | - | 6 | 846 | ||
AK145112.1 | - | 6 | 860 | |||
BC056365.1 | - | 6 | 857 | 6824847 | ||
BC058338.1 | - | 5 | 690 |