Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290502.1 | NP_001277431.1 | 5, 6 | 1040 | ||
NM_025978.4 | NP_080254.2 | 5, 6 | 802 | |||
NM_027619.4 | NP_081895.2 | 5, 6 | 802 | |||
GenBank mRNA | AK012246.1 | - | 5, 6 | 764 | ||
AK015308.1 | - | 5, 6 | 762 | |||
AK017763.1 | - | 3, 4 | 400 | |||
AK031605.1 | - | 5, 6 | 1033 | |||
AK035570.1 | - | 5, 6 | 772 | |||
AK041095.1 | - | 5, 6 | 776 | |||
AK044266.1 | - | 5, 6 | 777 | |||
BC021448.1 | - | 5, 6 | 810 | |||
BC024847.1 | - | 5, 6 | 767 | 5355806 | ||
BC138212.1 | - | 5, 6 | 764 | 8861231 | ||
BC145944.1 | - | 5, 6 | 764 | 40131247 |