Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026555.2 | NP_080831.2 | 2 | 330 | ||
GenBank mRNA | AK077943.1 | - | 2 | 270 | ||
AK082762.1 | - | 2 | 260 | |||
AK086705.1 | - | 2 | 270 | |||
AK132143.1 | - | 2 | 262 | |||
BC005487.1 | - | 1 | 167 | |||
BC025602.1 | - | 2 | 281 | 5250505 | ||
BC055903.1 | - | 2 | 277 | 4010685 | ||
BC092069.1 | - | 2 | 228 | 4215821 | ||
BY248337.1 | - | 2 | 332 |