Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030098.2 | NP_084374.2 | 2 | 880 | ||
GenBank mRNA | AK020595.1 | - | 2 | 823 | ||
AK038708.1 | - | 3 | 1196 | |||
AY545655.1 | - | 2 | 858 | |||
BC094892.1 | - | 2 | 835 | 6432787 |