Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290699.1 | NP_001277628.1 | 6, 7 | 849 | ||
NM_133817.3 | NP_598578.1 | 8, 9 | 1087 | |||
GenBank mRNA | AK030088.1 | - | 8, 9 | 920 | ||
BC062154.1 | - | 6, 7 | 639 | 6331597 |