Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135255.1 | NP_001128727.1 | 3 | 336 | ||
NM_001135256.1 | NP_001128728.1 | 3 | 239 | |||
NM_005610.2 | NP_005601.1 | 3 | 339 | |||
GenBank mRNA | AK222779.1 | - | 3 | 242 | ||
AK299251.1 | - | 3 | 239 | |||
AK312571.1 | - | 3 | 277 | |||
BC003092.1 | - | 3 | 266 | 3356558 | ||
BC053904.1 | - | 3 | 252 | 6526200 | ||
BC075836.1 | - | 3 | 207 | 6338602 | ||
BT007309.1 | - | 3 | 180 | |||
DC315298.1 | - | 3 | 340 | |||
X71810.1 | - | 3 | 253 | |||
X74262.1 | - | 3 | 264 |