Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trafd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163470.1 | NP_001156942.1 | 5 | 594 | ||
NM_172275.2 | NP_758479.2 | 5 | 594 | |||
GenBank mRNA | AK150038.1 | - | 5 | 563 | ||
BC038396.1 | - | 5 | 550 | 3987227 | ||
BU613615.1 | - | 5 | 594 |