Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005633.3 | NP_005624.2 | 2, 3 | 262 | ||
XM_005264515.4 | XP_005264572.1 | 2, 3 | 491 | |||
XM_011533062.2 | XP_011531364.1 | 2, 3 | 430 | |||
XM_011533064.2 | XP_011531366.1 | 2, 3 | 170 | |||
GenBank mRNA | AK290228.1 | - | 3, 4 | 308 | ||
AK301960.1 | - | 2, 3 | 321 | |||
L13857.1 | - | 2, 3 | 221 |