Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163701.1 | NP_001157173.1 | 25 | 4432 | ||
NM_177294.5 | NP_796268.3 | 26 | 4509 | |||
GenBank mRNA | AK004498.1 | - | 3 | 317 | ||
AK122505.1 | - | 26 | 4476 | |||
AK154007.1 | - | 25 | 4416 | |||
AK166232.1 | - | 25 | 4415 | |||
AK167589.1 | - | 25 | 4415 | |||
AK171233.1 | - | 24 | 3772 | |||
BC012218.1 | - | 4 | 912 | |||
BC051680.1 | - | 12 | 2425 | |||
BC088899.1 | - | 24 | 4237 | |||
BC117973.1 | - | 25 | 4401 | 40098643 | ||
BC117974.1 | - | 25 | 4401 | 40098644 |