Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199310.2 | NP_955014.2 | 4 | 975 | ||
GenBank mRNA | AK132604.1 | - | 4 | 967 | ||
AK133813.1 | - | 4 | 950 | |||
AK147152.1 | - | 4 | 951 | |||
AK152091.1 | - | 1 | 328 | |||
BC052687.1 | - | 4 | 942 |