Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205820.1 | NP_991389.1 | 3 | 966 | ||
GenBank mRNA | AK155696.1 | - | 3 | 947 | ||
AK170044.1 | - | 3 | 968 | |||
AY510706.1 | - | 3 | 966 | |||
BC117913.2 | - | 2 | 924 | 40097310 | ||
BC117914.2 | - | 2 | 924 | 40097311 |