Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164056.1 | NP_001157528.1 | 3 | 329 | ||
NM_008875.4 | NP_032901.2 | 2 | 274 | |||
GenBank mRNA | BC068144.1 | - | 2 | 131 | 30532470 | |
CF727963.1 | - | 2 | 274 | |||
U87868.1 | - | 3 | 312 |