Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164056.1 | NP_001157528.1 | 24 | 2855 | ||
NM_008875.4 | NP_032901.2 | 23 | 2800 | |||
GenBank mRNA | AK136111.1 | - | 18 | 2038 | ||
BC026625.1 | - | 4 | 315 | |||
BC068144.1 | - | 23 | 2657 | 30532470 | ||
U87868.1 | - | 24 | 2838 |