Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010452.3 | NP_034582.1 | 4 | 793 | ||
GenBank mRNA | BC096612.1 | - | 3 | 769 | 6476230 | |
U56399.1 | - | 1 | 302 | |||
Y11717.1 | - | 4 | 843 |