Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204466.1 | NP_001191395.1 | 9 | 1175 | ||
NM_001204467.1 | NP_001191396.1 | 10 | 1406 | |||
NM_001204468.1 | NP_001191397.1 | 10 | 1437 | |||
NM_005676.4 | NP_005667.2 | 10 | 1406 | |||
NM_152856.2 | NP_690595.1 | 9 | 1175 | |||
XM_005272677.4 | XP_005272734.1 | 10 | 1459 | |||
XM_005272678.4 | XP_005272735.1 | 9 | 990 | |||
XM_005272679.4 | XP_005272736.1 | 9 | 990 | |||
XM_006724563.1 | XP_006724626.1 | 6 | 623 | |||
XM_011543989.2 | XP_011542291.1 | 2 | 347 | |||
XM_017029884.1 | XP_016885373.1 | 9 | 1326 | |||
XM_017029885.1 | XP_016885374.1 | 9 | 1101 | |||
GenBank mRNA | AK225175.1 | - | 10 | 1365 | ||
AK292758.1 | - | 9 | 1126 | |||
BC000681.2 | - | 7 | 631 | |||
BC003089.1 | - | 9 | 1114 | 3536058 | ||
BC004181.2 | - | 10 | 1359 | 2961094 | ||
BC008733.2 | - | 10 | 1391 | 3163064 | ||
BC024153.2 | - | 9 | 1021 | 3845258 | ||
BX537969.1 | - | 10 | 1388 | |||
D50912.1 | - | 10 | 1257 | |||
KU178411.1 | - | 8 | 782 | |||
KU178412.1 | - | 9 | 1013 |