Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164661.1 | NP_001158133.1 | 32 | 3944 | ||
NM_001164662.1 | NP_001158134.1 | 31 | 3885 | |||
NM_011370.3 | NP_035500.2 | 31 | 3891 | |||
GenBank mRNA | AF072697.1 | - | 31 | 3686 | ||
AJ567911.1 | - | 30 | 3653 | |||
AK028811.1 | - | 22 | 2683 | |||
AK128952.1 | - | 31 | 3678 | |||
AK146613.1 | - | 31 | 3694 | |||
AK152558.1 | - | 32 | 3760 | |||
AK157773.1 | - | 31 | 3679 | |||
AK167068.1 | - | 31 | 3715 | |||
AK170572.1 | - | 1 | 1170 | |||
AK220320.1 | - | 31 | 3732 | |||
BC002174.1 | - | 3 | 317 | |||
BC047135.1 | - | 31 | 3680 | 6308554 | ||
BC052713.1 | - | 31 | 3750 | 6835403 | ||
BC054429.1 | - | 30 | 3664 | 5345535 | ||
BC086625.1 | - | 15 | 4666 |