Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL18BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039659.1 | NP_001034748.1 | 7 | 865 | ||
NM_001039660.1 | NP_001034749.1 | 6 | 753 | |||
NM_001145055.1 | NP_001138527.1 | 3 | 373 | |||
NM_001145057.1 | NP_001138529.1 | 6 | 691 | |||
NM_005699.3 | NP_005690.2 | 4 | 1740 | |||
NM_173042.2 | NP_766630.2 | 5 | 1509 | |||
NM_173044.2 | NP_766632.2 | 6 | 625 | |||
XM_017017059.1 | XP_016872548.1 | 6 | 1455 | |||
XM_017017060.1 | XP_016872549.1 | 6 | 1422 | |||
XM_017017061.1 | XP_016872550.1 | 7 | 1318 | |||
XM_017017062.1 | XP_016872551.1 | 7 | 1191 | |||
XM_017017063.1 | XP_016872552.1 | 7 | 1767 | |||
GenBank mRNA | AF110799.1 | - | 5 | 646 | ||
AF110800.1 | - | 3 | 374 | |||
AF110801.1 | - | 4 | 1740 | |||
AF122906.1 | - | 6 | 665 | |||
AF215907.2 | - | 6 | 614 | |||
AK098255.1 | - | 5 | 2603 | |||
BC044215.1 | - | 7 | 847 | 5755934 |