Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6pc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289856.1 | NP_001276785.1 | 5 | 1412 | ||
NM_001289857.1 | NP_001276786.1 | 4 | 1516 | |||
NM_021331.4 | NP_067306.1 | 5 | 1632 | |||
GenBank mRNA | AK148450.1 | - | 4 | 1517 | ||
AK148465.1 | - | 5 | 1584 | |||
BC111905.1 | - | 4 | 1491 | |||
Z47787.1 | - | 5 | 1610 |